siRNA Details
Gene Symbol: CERKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030311.2 | NP_001025482.1 | 4 | 760 | ||
NM_001160277.1 | NP_001153749.1 | 3 | 628 | |||
NM_201548.4 | NP_963842.1 | 4 | 760 | |||
GenBank mRNA | AJ640141.1 | - | 4 | 677 | ||
AJ697855.1 | - | 4 | 659 | |||
AK293844.1 | - | 3 | 629 | |||
AY357073.1 | - | 4 | 659 | |||
AY690329.1 | - | 4 | 659 | |||
BC137498.1 | - | 4 | 686 | 9021498 | ||
BC137499.1 | - | 4 | 686 | 9021499 | ||
HQ426666.1 | - | 4 | 566 | |||
HQ426671.1 | - | 4 | 488 | |||
HQ426672.1 | - | 4 | 488 | |||
HQ426677.1 | - | 2 | 96 | |||
HQ426678.1 | - | 5 | 813 | |||
HQ426679.1 | - | 3 | 421 | |||
JF290420.1 | - | 4 | 646 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
