siRNA Details
Gene Symbol: GOLGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004486.4 | NP_004477.3 | 11, 12 | 872 | ||
XM_005251930.1 | XP_005251987.1 | 12, 13 | 971 | |||
XM_005251931.1 | XP_005251988.1 | 11, 12 | 890 | |||
XM_005251932.1 | XP_005251989.1 | 10, 11 | 802 | |||
GenBank mRNA | AF248953.1 | - | 11, 12 | 823 | ||
BC069268.1 | - | 11, 12 | 872 | 6340972 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
