siRNA Details
Gene Symbol: ZNF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006959.2 | NP_008890.2 | 3 | 563 | ||
XM_017027205.1 | XP_016882694.1 | 4 | 2513 | |||
GenBank mRNA | AB075827.1 | - | 1 | 170 | ||
AK098183.1 | - | 3 | 545 | |||
AK127948.1 | - | 4 | 602 | |||
AK128471.1 | - | 1 | 768 |
