siRNA Details
Gene Symbol: LLGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031803.1 | NP_001026973.1 | 15, 16 | 2025 | ||
NM_004524.2 | NP_004515.2 | 15, 16 | 2025 | |||
XM_011524801.1 | XP_011523103.1 | 15, 16 | 2008 | |||
XM_011524802.1 | XP_011523104.1 | 15, 16 | 1996 | |||
XM_011524807.2 | XP_011523109.1 | 15, 16 | 2113 | |||
XM_017024625.1 | XP_016880114.1 | 15, 16 | 1996 | |||
XM_017024626.1 | XP_016880115.1 | 15, 16 | 1996 | |||
XM_017024627.1 | XP_016880116.1 | 15, 16 | 1996 | |||
XM_017024628.1 | XP_016880117.1 | 15, 16 | 1996 | |||
XM_017024629.1 | XP_016880118.1 | 15, 16 | 1996 | |||
XM_017024630.1 | XP_016880119.1 | 15, 16 | 1996 | |||
XM_017024631.1 | XP_016880120.1 | 15, 16 | 1996 | |||
XM_017024632.1 | XP_016880121.1 | 5, 6 | 1380 | |||
GenBank mRNA | AK025401.1 | - | 5, 6 | 1391 | ||
AK126687.1 | - | 12, 13 | 2048 | |||
AK301204.1 | - | 7, 8 | 862 | |||
AK315126.1 | - | 15, 16 | 1975 | |||
BC064994.1 | - | 15, 16 | 1998 | 5575350 | ||
X87342.1 | - | 15, 16 | 2001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
