siRNA Details
Gene Symbol: ERCC6L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020207.4 | NP_064592.2 | 19 | 4655 | ||
XM_011518641.2 | XP_011516943.1 | 18 | 4502 | |||
XM_011518644.2 | XP_011516946.1 | 18 | 4447 | |||
XM_011518648.2 | XP_011516950.1 | 10 | 2727 | |||
XM_011518650.2 | XP_011516952.1 | 19 | 3974 | |||
XM_017014707.1 | XP_016870196.1 | 19 | 4642 | |||
GenBank mRNA | AK095025.1 | - | 1 | 465 | ||
BC022957.1 | - | 4 | 1084 | 4995030 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
