siRNA Details
Gene Symbol: MAVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206491.1 | NP_001193420.1 | 6 | 3744 | ||
NM_020746.4 | NP_065797.2 | 7 | 3919 | |||
NR_037921.1 | 6 | 3746 | ||||
GenBank mRNA | AB033097.1 | - | 6 | 3825 | ||
AB097003.1 | - | 8 | 3283 | |||
AK023799.1 | - | 7 | 3874 | |||
AK123956.1 | - | 4 | 1741 | |||
BX649084.1 | - | 6 | 3704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
