siRNA Details
Gene Symbol: MARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039469.2 | NP_001034558.2 | 2 | 770 | ||
NM_001163296.1 | NP_001156768.1 | 2 | 770 | |||
NM_001163297.1 | NP_001156769.1 | 2 | 770 | |||
NM_004954.4 | NP_004945.4 | 2 | 770 | |||
NM_017490.3 | NP_059672.2 | 2 | 587 | |||
XM_011544789.1 | XP_011543091.1 | 3 | 661 | |||
XM_011544790.1 | XP_011543092.1 | 3 | 661 | |||
XM_011544791.1 | XP_011543093.1 | 3 | 661 | |||
XM_011544792.1 | XP_011543094.1 | 3 | 661 | |||
XM_011544793.1 | XP_011543095.1 | 3 | 661 | |||
XM_011544794.1 | XP_011543096.1 | 3 | 661 | |||
XM_011544795.1 | XP_011543097.1 | 3 | 661 | |||
XM_011544796.1 | XP_011543098.1 | 3 | 661 | |||
XM_011544797.1 | XP_011543099.1 | 3 | 661 | |||
XM_011544798.1 | XP_011543100.1 | 3 | 661 | |||
XM_017017310.1 | XP_016872799.1 | 2 | 743 | |||
XM_017017311.1 | XP_016872800.1 | 3 | 661 | |||
GenBank mRNA | AB188493.1 | - | 2 | 774 | ||
AB271694.1 | - | 1 | 92 | |||
AF387638.1 | - | 1 | 110 | |||
AK225766.1 | - | 2 | 605 | |||
AK290339.1 | - | 2 | 555 | |||
AK307858.1 | - | 3 | 406 | |||
BC008771.2 | - | 2 | 162 | |||
BC084540.1 | - | 2 | 229 | 4591688 | ||
BM724555.1 | - | 2 | 497 | |||
BT007342.1 | - | 1 | 92 | |||
BX383836.2 | - | 2 | 751 | |||
X97630.1 | - | 2 | 499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
