siRNA Details
Gene Symbol: ZNF814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144989.1 | NP_001138461.1 | 3 | 759 | ||
XM_017027197.1 | XP_016882686.1 | 2 | 404 | |||
GenBank mRNA | AK300483.1 | - | 3 | 722 |
Gene Symbol: ZNF552 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024762.3 | NP_079038.2 | 3 | 654 | ||
XM_005259267.3 | XP_005259324.1 | 3 | 832 | |||
GenBank mRNA | AK097041.1 | - | 3 | 654 | ||
AK293445.1 | - | 3 | 833 | |||
BC017460.1 | - | 1 | 92 | |||
BC046237.1 | - | 1 | 92 | |||
BC062982.1 | - | 1 | 92 | |||
BC082767.1 | - | 1 | 92 |
Gene Symbol: ZNF587B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204818.1 | NP_001191747.1 | 3 | 722 | ||
GenBank mRNA | AK299091.1 | - | 3 | 703 | ||
U71363.1 | - | 4 | 716 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
