siRNA Details
Gene Symbol: AKT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243027.2 | NP_001229956.1 | 5, 6 | 696 | ||
NM_001243028.2 | NP_001229957.1 | 4, 5 | 603 | |||
NM_001626.5 | NP_001617.1 | 5, 6 | 733 | |||
XM_011526614.1 | XP_011524916.1 | 5, 6 | 536 | |||
XM_011526615.1 | XP_011524917.1 | 5, 6 | 613 | |||
XM_011526616.1 | XP_011524918.1 | 5, 6 | 856 | |||
XM_011526618.1 | XP_011524920.1 | 5, 6 | 698 | |||
XM_011526619.1 | XP_011524921.1 | 5, 6 | 667 | |||
XM_011526620.1 | XP_011524922.1 | 5, 6 | 602 | |||
XM_011526622.2 | XP_011524924.1 | 5, 6 | 680 | |||
XM_017026470.1 | XP_016881959.1 | 6, 7 | 815 | |||
XM_017026471.1 | XP_016881960.1 | 5, 6 | 680 | |||
GenBank mRNA | AK055779.1 | - | 1, 2 | 762 | ||
AK294453.1 | - | 5, 6 | 643 | |||
AK304209.1 | - | 5, 6 | 742 | |||
AK308006.1 | - | 4, 5 | 472 | |||
AK308906.1 | - | 4, 5 | 550 | |||
AK314619.1 | - | 5, 6 | 675 | |||
BC032709.1 | - | 5, 6 | 587 | |||
BC040028.1 | - | 5, 6 | 698 | |||
BC120994.1 | - | 4, 5 | 479 | 40116490 | ||
BC120995.1 | - | 4, 5 | 479 | 40116493 | ||
KU159425.1 | - | 5, 6 | 680 | |||
M77198.1 | - | 5, 6 | 637 | |||
M95936.1 | - | 4, 5 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
