siRNA Details
Gene Symbol: CALML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031733.2 | NP_001026903.2 | 4 | 3792 | ||
NM_001286694.1 | NP_001273623.1 | 4 | 3902 | |||
NM_001286695.1 | NP_001273624.1 | 3 | 3761 | |||
NM_033429.2 | NP_219501.2 | 5 | 3933 | |||
NR_104583.1 | 2 | 3572 | ||||
GenBank mRNA | BX640702.1 | - | 3 | 2950 | ||
BX640817.1 | - | 2 | 3573 |
Gene Symbol: PIAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA837658.1 | - | 2 | 86 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
