siRNA Details
Gene Symbol: TCP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004610.3 | NP_004601.3 | 5 | 688 | ||
XM_011536082.2 | XP_011534384.1 | 3 | 584 | |||
XM_011536084.2 | XP_011534386.1 | 5 | 683 | |||
XM_011536085.2 | XP_011534387.1 | 5 | 862 | |||
XM_011536088.2 | XP_011534390.1 | 4 | 502 | |||
XM_011536090.2 | XP_011534392.1 | 4 | 449 | |||
XM_017011228.1 | XP_016866717.1 | 4 | 542 | |||
XM_017011229.1 | XP_016866718.1 | 4 | 542 | |||
XM_017011230.1 | XP_016866719.1 | 4 | 542 | |||
XM_017011231.1 | XP_016866720.1 | 4 | 542 | |||
GenBank mRNA | BC063451.1 | - | 5 | 644 | 5170229 | |
U03399.1 | - | 5 | 634 |
Gene Symbol: TCP10L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145121.1 | NP_001138593.1 | 5 | 688 | ||
XM_017010859.1 | XP_016866348.1 | 5 | 659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
