siRNA Details
Gene Symbol: TAAR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053278.1 | NP_444508.1 | 1 | 793 | ||
GenBank mRNA | AY183468.1 | - | 1 | 793 | ||
BC069166.1 | - | 1 | 793 | 7217002 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TAAR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053278.1 | NP_444508.1 | 1 | 793 | ||
GenBank mRNA | AY183468.1 | - | 1 | 793 | ||
BC069166.1 | - | 1 | 793 | 7217002 |