siRNA Details
Gene Symbol: TOP3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320759.1 | NP_001307688.1 | 4 | 705 | ||
NM_004618.4 | NP_004609.1 | 5 | 779 | |||
XM_011524001.2 | XP_011522303.1 | 1 | 78 | |||
GenBank mRNA | AK126869.1 | - | 2 | 344 | ||
AK292926.1 | - | 5 | 602 | |||
AK296437.1 | - | 4 | 537 | |||
AK311668.1 | - | 3 | 546 | |||
BC051748.1 | - | 5 | 661 | 6044661 | ||
BM462184.1 | - | 780 | ||||
U43431.1 | - | 5 | 642 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
