siRNA Details
Gene Symbol: SGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012410.4 | NP_001012410.1 | 9 | 2072 | ||
NM_001012412.4 | NP_001012412.1 | 9 | 1316 | |||
NM_001199252.2 | NP_001186181.1 | 9 | 2183 | |||
NM_001199254.2 | NP_001186183.1 | 9 | 1427 | |||
NM_001199256.2 | NP_001186185.1 | 8 | 1376 | |||
NM_001199257.2 | NP_001186186.1 | 7 | 1145 | |||
NM_138484.4 | NP_612493.1 | 8 | 1265 | |||
GenBank mRNA | AB187578.1 | - | 9 | 2030 | ||
AB187580.1 | - | 9 | 1274 | |||
AB187582.1 | - | 8 | 1223 | |||
BC017867.1 | - | 8 | 1223 | 4685140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
