siRNA Details
Gene Symbol: MRFAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203462.2 | NP_982287.1 | 2 | 1450 | ||
XM_005247932.1 | XP_005247989.1 | 2 | 1441 | |||
GenBank mRNA | AF155654.1 | - | 2 | 1445 | ||
AF258591.1 | - | 1 | 1985 | |||
BC008087.1 | - | 2 | 1397 | 3446352 | ||
BC066897.1 | - | 2 | 1449 | 4818881 |
