siRNA Details
Gene Symbol: FRMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018071.3 | NP_001018081.3 | 13 | 1809 | ||
NM_001318191.1 | NP_001305120.1 | 11 | 1450 | |||
XM_011539327.2 | XP_011537629.1 | 13 | 1607 | |||
GenBank mRNA | AK123038.1 | - | 11 | 1450 | ||
AK131386.1 | - | 11 | 1295 | |||
AK303923.1 | - | 13 | 1507 | |||
CR749241.1 | - | 13 | 1781 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
