siRNA Details
Gene Symbol: FRMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018071.3 | NP_001018081.3 | 16 | 2352 | ||
NM_001318191.1 | NP_001305120.1 | 14 | 1993 | |||
GenBank mRNA | AK123038.1 | - | 14 | 1993 | ||
AK131386.1 | - | 14 | 1838 | |||
AK303923.1 | - | 16 | 2050 | |||
CR749241.1 | - | 16 | 2324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
