siRNA Details
Gene Symbol: BTNL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017009116.1 | XP_016864605.1 | 1 | 503 | ||
XM_017009117.1 | XP_016864606.1 | 1 | 503 | |||
GenBank mRNA | AK057097.1 | - | 6 | 1573 | ||
AK298113.1 | - | 5 | 1412 | |||
AK309516.1 | - | 3 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
