siRNA Details
Gene Symbol: ARMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282745.1 | NP_001269674.1 | 13 | 1719 | ||
NM_001282746.1 | NP_001269675.1 | 13 | 1719 | |||
NM_001282747.1 | NP_001269676.1 | 10 | 1230 | |||
NM_173081.4 | NP_775104.2 | 13 | 1719 | |||
XM_005252380.2 | XP_005252437.1 | 12 | 1507 | |||
XM_005252381.2 | XP_005252438.1 | 12 | 1540 | |||
XM_011519350.2 | XP_011517652.1 | 13 | 1702 | |||
XM_011519351.2 | XP_011517653.1 | 4 | 549 | |||
XM_017015831.1 | XP_016871320.1 | 12 | 1507 | |||
XM_017015832.1 | XP_016871321.1 | 13 | 1702 | |||
XM_017015833.1 | XP_016871322.1 | 12 | 1507 | |||
GenBank mRNA | AK057389.1 | - | 13 | 1662 | ||
AK098711.1 | - | 3 | 259 | |||
AK302104.1 | - | 10 | 1177 | |||
AK302985.1 | - | 13 | 1706 | |||
AY436643.1 | - | 13 | 1680 | |||
BC039312.1 | - | 13 | 1712 | 5296302 |
