siRNA Details
Gene Symbol: ZNF621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098414.2 | NP_001091884.1 | 5 | 1899 | ||
NM_198484.4 | NP_940886.1 | 5 | 2135 | |||
XM_005265079.3 | XP_005265136.1 | 5 | 1962 | |||
XM_005265080.3 | XP_005265137.1 | 4 | 1881 | |||
XM_006713114.3 | XP_006713177.1 | 5 | 1843 | |||
XM_011533610.2 | XP_011531912.1 | 5 | 2469 | |||
XM_017006226.1 | XP_016861715.1 | 4 | 1762 | |||
GenBank mRNA | AK127181.1 | - | 5 | 1899 | ||
CR749268.1 | - | 5 | 2000 |
