siRNA Details
Gene Symbol: SPHK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204158.2 | NP_001191087.1 | 6 | 989 | ||
NM_001204159.2 | NP_001191088.1 | 6 | 1070 | |||
NM_001204160.2 | NP_001191089.1 | 5 | 886 | |||
NM_001243876.1 | NP_001230805.1 | 5 | 694 | |||
NM_020126.4 | NP_064511.2 | 6 | 1166 | |||
XM_006723292.1 | XP_006723355.1 | 5 | 856 | |||
XM_011527133.1 | XP_011525435.1 | 6 | 1001 | |||
XM_011527134.1 | XP_011525436.1 | 5 | 858 | |||
XM_017027008.1 | XP_016882497.1 | 4 | 1572 | |||
XM_017027009.1 | XP_016882498.1 | 5 | 577 | |||
XM_017027010.1 | XP_016882499.1 | 6 | 1056 | |||
GenBank mRNA | AB100373.1 | - | 4 | 328 | ||
AF245447.1 | - | 4 | 698 | |||
AK000599.1 | - | 5 | 800 | |||
AK074601.1 | - | 1639 | ||||
AK122735.1 | - | 7 | 870 | |||
AK297787.1 | - | 6 | 948 | |||
AK300541.1 | - | 6 | 828 | |||
AL136701.1 | - | 6 | 1069 | |||
BC006161.1 | - | 6 | 1166 | 3677636 | ||
BC010671.1 | - | 5 | 886 | 3870258 | ||
DA926908.1 | - | 5 | 533 | |||
DB053266.1 | - | 4 | 469 | |||
EF107108.1 | - | 4 | 1694 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
