siRNA Details
Gene Symbol: C9orf131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040410.2 | NP_001035500.1 | 2 | 1753 | ||
NM_001040411.2 | NP_001035501.1 | 2 | 1642 | |||
NM_001040412.2 | NP_001035502.1 | 3 | 1717 | |||
NM_001287391.1 | NP_001274320.1 | 2 | 1674 | |||
NM_203299.3 | NP_976044.2 | 2 | 1858 | |||
GenBank mRNA | AK097595.1 | - | 2 | 1824 | ||
AK302123.1 | - | 3 | 441 | |||
BC045643.1 | - | 2 | 1837 | 5298387 |
