siRNA Details
Gene Symbol: RAPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203365.3 | NP_976241.1 | 16 | 2344 | ||
GenBank mRNA | AY523978.1 | - | 16 | 2340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
