siRNA Details
Gene Symbol: XKRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004677.2 | NP_004668.2 | 1 | 1092 | ||
GenBank mRNA | AF000997.1 | - | 1 | 1091 | ||
BC132883.1 | - | 1 | 964 | 40146905 | ||
BC132885.1 | - | 1 | 964 | 40146907 |
Gene Symbol: XKRY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002906.1 | NP_001002906.1 | 1 | 1092 | ||
GenBank mRNA | BC132901.1 | - | 1 | 964 | 40146923 | |
BC132903.1 | - | 1 | 964 | 40146925 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
