siRNA Details
Gene Symbol: SPACA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205856.3 | NP_995328.2 | 3 | 458 | ||
XM_017029512.1 | XP_016885001.1 | 2 | 1073 | |||
GenBank mRNA | AF217622.1 | - | 2 | 314 | ||
AY358844.1 | - | 3 | 458 | |||
BX279755.1 | - | 2 | 307 |
Gene Symbol: SPACA5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079900.1 | NP_001073369.1 | 2 | 321 |

More Information
Other Species Targeted

Gene Ontology Categories:
