siRNA Details
Gene Symbol: TMEM132E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304438.1 | NP_001291367.1 | 9 | 3522 | ||
GenBank mRNA | BC018318.1 | - | 1 | 948 | ||
BC020591.1 | - | 7 | 2177 |

More Information
Other Species Targeted

