siRNA Details
Gene Symbol: ACSM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207321.2 | NP_997204.2 | 9, 10 | 1319 | ||
GenBank mRNA | AK126134.1 | - | 8, 9 | 1185 | ||
BC063654.1 | - | 6, 7 | 708 | |||
BC104210.1 | - | 8, 9 | 1203 | |||
BC104211.2 | - | 8, 9 | 1203 | |||
BC112988.1 | - | 9, 10 | 1284 | |||
BC148184.1 | - | 8, 9 | 1203 |
