siRNA Details
Gene Symbol: C2CD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207322.2 | NP_997205.2 | 2 | 1311 | ||
GenBank mRNA | AF504646.1 | - | 2 | 1285 | ||
BX648578.1 | - | 1 | 29 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
