siRNA Details
Gene Symbol: PIEZO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022068.3 | NP_071351.2 | 17 | 2612 | ||
XM_011525723.2 | XP_011524025.1 | 17 | 3410 | |||
XM_011525724.2 | XP_011524026.1 | 17 | 3410 | |||
XM_011525725.2 | XP_011524027.1 | 17 | 3410 | |||
XM_011525726.2 | XP_011524028.1 | 17 | 3410 | |||
XM_017025918.1 | XP_016881407.1 | 16 | 3371 | |||
GenBank mRNA | EG327967.1 | - | 3 | 300 | ||
JN790819.1 | - | 17 | 2446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
