siRNA Details
Gene Symbol: POTEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110752.1 | 8 | 1797 | |||
XM_011543796.2 | XP_011542098.1 | 7 | 1623 | |||
GenBank mRNA | BC066981.1 | - | 8 | 1797 | 4824630 |
Gene Symbol: LOC100288966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011545655.2 | XP_011543957.1 | 7 | 1353 |
Gene Symbol: LOC102723502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546199.2 | XP_011544501.1 | 7 | 1352 |
Gene Symbol: POTEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543794.2 | XP_011542096.1 | 7 | 1241 |
Gene Symbol: POTEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011543793.2 | XP_011542095.1 | 7 | 1241 |
Gene Symbol: POTED | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011529550.2 | XP_011527852.1 | 7 | 1353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
