siRNA Details
Gene Symbol: C1orf174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207356.2 | NP_997239.2 | 3 | 305 | ||
XM_005244743.3 | XP_005244800.1 | 4 | 601 | |||
XM_011541323.2 | XP_011539625.1 | 4 | 2793 | |||
GenBank mRNA | AK125092.1 | - | 3 | 1709 | ||
AK289493.1 | - | 3 | 305 | |||
BC035643.1 | - | 3 | 268 | |||
BC067302.1 | - | 3 | 297 | 30409047 |
