siRNA Details
Gene Symbol: LINC01140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026985.1 | 1 | 319 | |||
NR_026986.1 | 1 | 319 | ||||
GenBank mRNA | AL833352.1 | - | 1 | 242 | ||
BC041867.1 | - | 1 | 290 | |||
BC126240.1 | - | 1 | 248 | 8991956 | ||
BC132861.1 | - | 1 | 248 | |||
DA398164.1 | - | 1 | 306 | |||
DB462107.2 | - | 1 | 322 |
