siRNA Details
Gene Symbol: NCKAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207363.2 | NP_997246.2 | 15 | 5402 | ||
XM_005263659.3 | XP_005263716.1 | 14 | 5984 | |||
XM_005263660.3 | XP_005263717.1 | 14 | 5499 | |||
XM_011511097.2 | XP_011509399.1 | 16 | 6172 | |||
XM_011511098.2 | XP_011509400.1 | 16 | 6169 | |||
XM_011511099.2 | XP_011509401.1 | 15 | 5567 | |||
XM_011511100.2 | XP_011509402.1 | 15 | 5665 | |||
XM_011511101.1 | XP_011509403.1 | 13 | 5075 | |||
XM_011511102.2 | XP_011509404.1 | 14 | 9501 | |||
XM_011511103.2 | XP_011509405.1 | 12 | 5303 | |||
XM_011511104.1 | XP_011509406.1 | 12 | 4954 | |||
XM_011511105.2 | XP_011509407.1 | 12 | 5745 | |||
XM_017003974.1 | XP_016859463.1 | 15 | 5295 | |||
XM_017003975.1 | XP_016859464.1 | 14 | 5597 | |||
XM_017003976.1 | XP_016859465.1 | 16 | 6193 | |||
XM_017003977.1 | XP_016859466.1 | 13 | 6103 | |||
XM_017003978.1 | XP_016859467.1 | 9 | 5513 | |||
GenBank mRNA | AK124659.1 | - | 2 | 274 | ||
AY946008.1 | - | 10 | 4824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
