siRNA Details
Gene Symbol: LRRC37A8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126038.1 | 1 | 1933 | |||
GenBank mRNA | AK125932.1 | - | 1 | 859 | ||
AK128235.1 | - | 1 | 1933 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
