siRNA Details
Gene Symbol: TGM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323316.1 | NP_001310245.1 | 14 | 2160 | ||
NM_001323317.1 | NP_001310246.1 | 12 | 1934 | |||
NM_001323318.1 | NP_001310247.1 | 12 | 1997 | |||
NM_004613.3 | NP_004604.2 | 13 | 2177 | |||
XM_011529028.1 | XP_011527330.1 | 14 | 2122 | |||
GenBank mRNA | AK126508.1 | - | 3 | 1591 | ||
AK291714.1 | - | 13 | 2018 | |||
AK295775.1 | - | 12 | 1775 | |||
AK300292.1 | - | 12 | 1838 | |||
AK314618.1 | - | 13 | 2018 | |||
AY675221.1 | - | 12 | 1946 | |||
BC073946.1 | - | 2 | 165 | |||
BQ430537.1 | - | 2 | 161 | |||
EU794665.1 | - | 13 | 2046 | |||
M55153.1 | - | 13 | 2081 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
