siRNA Details
Gene Symbol: LINC00243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130726.1 | 2 | 1434 | |||
GenBank mRNA | AK098012.1 | - | 2 | 1434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
