siRNA Details
Gene Symbol: C7orf76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201450.1 | NP_001188379.1 | 5 | 582 | ||
NM_001201451.1 | NP_001188380.1 | 4 | 552 | |||
GenBank mRNA | AK124274.1 | - | 5 | 582 | ||
BC140736.1 | - | 4 | 539 | 9021604 | ||
BC171740.1 | - | 4 | 539 | 9052960 |
