siRNA Details
Gene Symbol: FAM74A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026801.1 | 2 | 1544 |
Gene Symbol: FAM74A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026802.1 | 2 | 1521 | |||
NR_110998.1 | 3 | 1605 | ||||
GenBank mRNA | AK127145.1 | - | 2 | 1372 |
Gene Symbol: FAM74A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126166.1 | 2 | 1554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
