siRNA Details
Gene Symbol: TOP3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320759.1 | NP_001307688.1 | 4, 5 | 743 | ||
NM_004618.4 | NP_004609.1 | 5, 6 | 817 | |||
XM_011524001.2 | XP_011522303.1 | 1, 2 | 116 | |||
GenBank mRNA | AK126869.1 | - | 2, 3 | 382 | ||
AK292926.1 | - | 5, 6 | 640 | |||
AK296437.1 | - | 4, 5 | 575 | |||
AK311668.1 | - | 3, 4 | 584 | |||
BC051748.1 | - | 5, 6 | 699 | 6044661 | ||
U43431.1 | - | 5, 6 | 680 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
