siRNA Details
Gene Symbol: STK31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260504.1 | NP_001247433.1 | 4 | 571 | ||
NM_001260505.1 | NP_001247434.1 | 4 | 295 | |||
NM_031414.4 | NP_113602.2 | 4 | 295 | |||
NM_032944.3 | NP_116562.2 | 4 | 539 | |||
NR_048542.1 | 4 | 287 | ||||
XM_006715758.3 | XP_006715821.1 | 4 | 209 | |||
XM_011515449.2 | XP_011513751.2 | 4 | 221 | |||
XM_011515450.1 | XP_011513752.1 | 4 | 542 | |||
XM_011515452.2 | XP_011513754.2 | 4 | 222 | |||
GenBank mRNA | AF285599.1 | - | 4 | 191 | ||
AF332194.1 | - | 4 | 574 | |||
AK057271.1 | - | 4 | 243 | |||
AK292474.1 | - | 4 | 187 | |||
AK302689.1 | - | 4 | 187 | |||
BC033018.1 | - | 4 | 189 | |||
BC059374.1 | - | 4 | 202 | 5268051 | ||
DB065148.1 | - | 4 | 243 | |||
DB086591.1 | - | 4 | 298 | |||
DB089624.1 | - | 4 | 199 | |||
DB095141.1 | - | 4 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
