siRNA Details
Gene Symbol: BRSK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032430.1 | NP_115806.1 | 1, 2 | 412 | ||
XM_005259327.3 | XP_005259384.1 | 1, 2 | 477 | |||
GenBank mRNA | AF479826.1 | - | 3, 4 | 310 | ||
AF479827.1 | - | 1, 2 | 412 | |||
AL834275.1 | - | 1, 2 | 35 | |||
AY458602.1 | - | 1, 2 | 184 | |||
AY505124.1 | - | 1, 2 | 135 | |||
HQ830199.1 | - | 1, 2 | 157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
