siRNA Details
Gene Symbol: Pten | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008960.2 | NP_032986.1 | 8 | 1845 | ||
GenBank mRNA | AK030750.1 | - | 4 | 608 | ||
AK076980.1 | - | 8 | 1803 | |||
AK088717.1 | - | 8 | 1824 | |||
AK138025.1 | - | 1 | 154 | |||
AK148736.1 | - | 8 | 1680 | |||
BC021445.1 | - | 8 | 1194 | 5038272 | ||
U92437.1 | - | 8 | 1925 |
