siRNA Details
Gene Symbol: Ptpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127177.1 | NP_001120649.1 | 2, 3 | 274 | ||
NM_008977.3 | NP_033003.1 | 2, 3 | 274 | |||
GenBank mRNA | AK014528.1 | - | 2, 3 | 242 | ||
AK076072.1 | - | 2, 3 | 227 | |||
AK132013.1 | - | 2, 3 | 237 | |||
BC008269.1 | - | 2, 3 | 248 | 3602983 | ||
CA560046.1 | - | 2, 3 | 274 | |||
M80739.1 | - | 2, 3 | 229 | |||
M81477.1 | - | 2, 3 | 229 | |||
S52655.1 | - | 2, 3 | 224 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053990, XM_008772080, XM_006254854, XM_006254855, XM_008772081, and others.
Gene Ontology Categories:
