siRNA Details
Gene Symbol: STK19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004197.1 | NP_004188.1 | 1 | 232 | ||
NM_032454.1 | NP_115830.1 | 1 | 232 | |||
NR_026717.1 | 2 | 332 | ||||
GenBank mRNA | AK302111.1 | - | 2 | 346 | ||
AK303438.1 | - | 1 | 270 | |||
BC110347.1 | - | 2 | 332 | 5165123 | ||
BC121128.1 | - | 1 | 166 | 40120738 | ||
BC121129.1 | - | 1 | 165 | 40120740 | ||
BC143833.1 | - | 1 | 202 | 9052348 | ||
BC143835.1 | - | 1 | 202 | |||
L26260.1 | - | 1 | 232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
