siRNA Details
Gene Symbol: PCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291556.1 | NP_001278485.1 | 9, 10 | 1585 | ||
NM_001308054.1 | NP_001294983.1 | 9, 10 | 2012 | |||
NM_004563.3 | NP_004554.3 | 9, 10 | 1751 | |||
GenBank mRNA | AK129934.1 | - | 9, 10 | 1569 | ||
AK301400.1 | - | 9, 10 | 1595 | |||
AK316206.1 | - | 9, 10 | 1405 | |||
BC001454.2 | - | 9, 10 | 1541 | 3138368 | ||
CR456913.1 | - | 9, 10 | 1476 | |||
KU178243.1 | - | 9, 10 | 1476 | |||
KU178245.1 | - | 3, 4 | 165 | |||
X92720.1 | - | 9, 10 | 1542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
