siRNA Details
Gene Symbol: Atxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199304.1 | NP_001186233.1 | 6 | 1762 | ||
NM_001199305.1 | NP_001186234.1 | 7 | 1887 | |||
NM_009124.6 | NP_033150.2 | 7 | 1887 | |||
GenBank mRNA | AK048268.1 | - | 7 | 1874 | ||
AK169931.1 | - | 7 | 1882 | |||
BC058178.1 | - | 6 | 1728 | 5326559 | ||
X83542.1 | - | 6 | 1628 |
