siRNA Details
Gene Symbol: RAB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271998.1 | NP_001258927.1 | 6 | 1645 | ||
NM_004578.3 | NP_004569.2 | 8 | 1879 | |||
NR_073545.1 | 8 | 1841 | ||||
GenBank mRNA | AY585832.1 | - | 8 | 1809 | ||
BC004309.1 | - | 8 | 1809 | 3627110 |
Gene Symbol: SPHAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006542.3 | NP_006533.1 | 1 | 356 | ||
GenBank mRNA | BC107681.1 | - | 1 | 356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
