siRNA Details
Gene Symbol: Sgca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009161.4 | NP_033187.1 | 5 | 580 | ||
GenBank mRNA | AB024920.1 | - | 5 | 580 | ||
AF019564.1 | - | 4 | 327 | |||
AK009015.1 | - | 4 | 343 | |||
AK075915.1 | - | 4 | 352 | |||
BC060287.1 | - | 4 | 339 |
