siRNA Details
Gene Symbol: TCP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011231.1 | XP_016866720.1 | 8 | 1126 | ||
GenBank mRNA | U03399.1 | - | 9 | 1215 |
Gene Symbol: TCP10L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010859.1 | XP_016866348.1 | 9 | 1243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
