siRNA Details
Gene Symbol: Hyal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028957.2 | NP_083233.1 | 3 | 1268 | ||
GenBank mRNA | AK017112.1 | - | 3 | 1283 | ||
AK133387.1 | - | 3 | 1271 | |||
BC145992.1 | - | 3 | 1201 | 40131295 | ||
BC145994.1 | - | 3 | 1201 | 40131297 |
Gene Symbol: Spam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079875.2 | NP_001073344.1 | 4 | 1296 | ||
NM_009241.3 | NP_033267.2 | 3 | 1290 | |||
GenBank mRNA | AK005638.1 | - | 4 | 1283 | ||
AY228460.1 | - | 2 | 971 | |||
AY920282.1 | - | 3 | 1290 | |||
AY920283.1 | - | 4 | 1269 | |||
U33958.1 | - | 3 | 1280 |

More Information
Other Species Targeted

Gene Ontology Categories:
